Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ BIOshell™ A160 Peptide Phenyl-Hexyl HPLC Column, 2.7 μm Particle Size

Catalog No. 111022303 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
2 cm
3 cm
5 cm
7.5 cm
10 cm
15 cm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-102-2303 15 cm 4.6 mm
11-102-2286 2 cm 2.1 mm
11-102-2287 3 cm 2.1 mm
11-102-2288 5 cm 2.1 mm
11-102-2296 10 cm 3 mm
11-102-2297 15 cm 3 mm
11-102-2298 2 cm 4.6 mm
11-102-2299 3 cm 4.6 mm
11-102-2300 5 cm 4.6 mm
11-102-2301 7.5 cm 4.6 mm
11-102-2302 10 cm 4.6 mm
11-102-2289 7.5 cm 2.1 mm
11-102-2290 10 cm 2.1 mm
11-102-2291 15 cm 2.1 mm
11-102-2292 2 cm 3 mm
11-102-2293 3 cm 3 mm
11-102-2294 5 cm 3 mm
11-102-2295 7.5 cm 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 11-102-2303 Supplier MilliporeSigma™ Supelco™ Supplier No. 577540U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A high-speed, high-performance liquid chromatography column based on a 160 Å Fused-Core™ particle design.

  • Particle provides a thin, porous shell of high-purity silica surrounding a solid silica core
  • Particle design exhibits very high column efficiency for high molecular weight solutes (≤ 20 kDa) due to the shallow diffusion paths in the 0.5 μm thick porous shell, and the small overall particle size of 2.7 μm
  • The end-capped, phenyl-hexyl bonded phase of the peptide phenyl-hexyl provides a stable, reversed phase packing
  • Optimized for reversed-phase HPLC separations of peptides and polypeptides, using typical acidic mobile phases favored for proteomic applications
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 15 cm
Particle Size 2.7 μm
pH 2 to 9
Pore Size 160 Å
Surface Area 90 m2/g
USP Type L11
Stationary Phase Phenyl-Hexyl (dimethylphenyl-hexylsilane)
Product Line BIOshell
Matrix Phenyl-Hexyl (dimethylphenyl-hexylsilane) Phase/Fused-Core™ Particle Platform
Volume (Metric) L11
Carbon Load 4.7%
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel
Technique Fused-Core HPLC
Show More Show Less

BIOshell is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.