Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ BIOshell™ IgG 1000 Å Diphenyl HPLC Guard Cartridge, 2.7 μm Particle Size

Catalog No. 111008394 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
5 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-100-8394 5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-8394 Supplier MilliporeSigma™ Supelco™ Supplier No. 577433U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BIOshell™ IgG 1,000 Å Diphenyl is a high-speed, high-performance liquid chromatography column based on a new wide-pore (1,000 Å) Fused-Core™ particle design.

BIOshell™ IgG 1,000 Å Diphenyl guard columns contain particle that provide a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5 μm thin porous shell and the small overall particle size of 2.7 μm. The densely bonded, extensively endcapped diphenylmethyl bonded phase of BIOshell™ IgG 1,000 Å Diphenyl provides a stable, reversed-phase packing that can be used for separating high molecular weight compounds, such as proteins.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 5 cm
Particle Size 2.7 μm
pH 2 to 9
Pore Size 1000 Å
Quantity 3/Pk.
Surface Area 22 m2/g
Stationary Phase Diphenyl (diphenylmethyl)
Product Line BIOshell
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel
Technique HPLC, LC/MS, UHPLC, UHPLC-MS
Product Type Guard Cartridge
Show More Show Less

BIOshell is a trademark of Sigma-Aldrich Co. LLC. Fused-Core is a registered trademark of Advanced Materials Technology, Inc.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.