Learn More
ILC Dover BioShield™ Full Hood
Designed in collaboration with leading healthcare organizations, this loose-fitting PAPR with an APF of 1,000 is made from spun bonded polyolefin, coated, and latex free.
$694.60 - $5575.21
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
19-001-504
|
ILC Dover
S20283 |
3/Pk. |
Pack of 3 for $694.60
|
|
|||||
19-001-505
|
ILC Dover
S202825 |
25/Pk. |
Pack of 25 for $5,575.21
|
|
|||||
Description
- Visor is impact resistant, made with optical grade polycarbonate
- Internal and external skirt interfaces with most protective suits and requires no fit testing
Specifications
Polycarbonate, Polyolefin | |
Yes |