Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories BioSign™ Flu A/B Kit

Catalog No. 23111369
Change view
Click to view available options
Quantity:
25 Tests
6 x 25 Kits = 150 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-369 25 Tests
23-111-370 6 x 25 Kits = 150 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-111-369 Supplier Germaine™ Laboratories Supplier No. BSP51025
Add to Cart
Add to Cart
This item is not returnable. View return policy

An In vitro rapid qualitative test that detects influenza type A and type B antigens directly from nasal swab, nasopharyngeal swab and nasal aspirate/wash specimens.

  • Differentiates between Influenza Type A and B, on a single test device
  • Produces results in 10 to 15 minutes
  • Each test device includes built-in controls
  • Each kit comes with external controls
  • Meets FDA performance criteria for influenza tests stated in code 21 CFR 866.3328
  • CLIA Complexity: Moderate complexity when used with nasal wash/aspirate samples
  • CLIA Waived when used with nasal and nasopharyngeal swabs

Specifications

Content And Storage 2°C to 30°C
CE Marker Yes
Clia Complexity Waived (Nasal Swab)
Detectable Analytes Influenza A and B
Includes Positive and Negative Control Swabs
Quantity 25 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.