Learn More
Agilent BioTek BioSpa 8 Automated Incubator



The BioTek BioSpa live cell analysis system offers unique benefits for a wide variety of live cell imaging applications in up to 8 microplates or other labware. For Research Use Only. Not for use in diagnostic procedures.
Specifications
Operating Temperature Range (Metric) | Ambient to 45°C |
---|---|
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Depth (English) | 20.1 in. |
Depth (Metric) | 51 cm |
Height (English) | 18.9 in. |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
11-120-550
![]() ![]() |
Agilent BioTek
BIOSPASN |
8-plate capacity; Incubation to 45°C; BioSpa software; One alignment kit | N/A | N/A | |||||
This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or send email to cs.webemail@thermofisher.com to discuss your specific assay requirements. | |||||||||
11-120-551
![]() ![]() |
Agilent BioTek
BIOSPAGSN |
8-plate capacity; Incubation to 45°C; CO2/O2 control and monitoring; Water pan and humidity monitoring; BioSpa software; One alignment kit | N/A | N/A | |||||
This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or send email to cs.webemail@thermofisher.com to discuss your specific assay requirements. | |||||||||
Description
The BioTek BioSpa live cell analysis system offers unique benefits for a wide variety of live cell imaging applications in up to 8 microplates or other labware. With 5 imaging modes, the system enables a wide range of microscopy applications
Capture detailed cellular images, get cell-level analyses for migration and wound healing assays, 3D spheroid formation, and many other kinetic applications. Built-in scheduling, environmental monitoring, and available liquid handling for walk away automation. Multiple users can run simultaneous processes for maximum efficiency in the lab.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Ambient to 45°C | |
20.1 in. | |
18.9 in. | |
80 to 95% | |
69 cm |
CE and TUV marked. RoHS compliant. | |
51 cm | |
48 cm | |
27.2 in. | |
Automated Microplate Incubator |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.