Learn More
Agilent BioTek BioSpa 8 Automated Incubator, IVD

The BioTek BioSpa live cell analysis system offers unique benefits for a wide variety of live cell imaging applications in up to 8 microplates or other labware. For In Vitro Diagnostic Use.
Specifications
Operating Temperature Range (Metric) | Ambient to 45°C |
---|---|
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Depth (English) | 20.1 in. |
Depth (Metric) | 51 cm |
Height (English) | 18.9 in. |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
BTBIOSPASI
![]() |
Agilent BioTek
BIOSPASI |
8-plate capacity; Incubation to 45°C; BioSpa software; One alignment kit | N/A | N/A | |||||
This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or send email to cs.webemail@thermofisher.com to discuss your specific assay requirements. | |||||||||
BTBIOSPAGSI
![]() |
Agilent BioTek
BIOSPAGSI |
8-plate capacity; Incubation to 45°C; CO2/O2 control and monitoring; Water pan and humidity monitoring; BioSpa software; One alignment kit | N/A | N/A | |||||
This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or send email to cs.webemail@thermofisher.com to discuss your specific assay requirements. | |||||||||
Description
The BioTek BioSpa live cell analysis system offers unique benefits for a wide variety of live cell imaging applications in up to 8 microplates or other labware. With 5 imaging modes, the system enables a wide range of microscopy applications
Capture detailed cellular images, get cell-level analyses for migration and wound healing assays, 3D spheroid formation, and many other kinetic applications. Built-in scheduling, environmental monitoring, and available liquid handling for walk away automation. Multiple users can run simultaneous processes for maximum efficiency in the lab.
Specifications
Ambient to 45°C | |
20.1 in. | |
18.9 in. | |
80 to 95% | |
69 cm |
CE and TUV marked. RoHS compliant. | |
51 cm | |
48 cm | |
27.2 in. | |
Automated Microplate Incubator |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.