Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek BioStack Automated Microplate Stacker - Microplate Stacks
GSA_VAGREENER_CHOICE

Catalog No. 11120603 Shop All Agilent Technologies Inc. Products
Change view
Click to view available options
Includes:
2 stacks of 10 microplates
2 stacks of 30 microplates
2 stacks of 50 microplates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
11-120-603 2 stacks of 50 microplates
BT7310030 2 stacks of 10 microplates
BT7310008 2 stacks of 30 microplates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 11-120-603 Supplier Agilent BioTek Supplier No. 7310023
Add to Cart
Add to Cart
This item is not returnable. View return policy

Storage stacks

  • One set

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Microplates
For Use With (Equipment) Bio-tek Biostack automated microplate stacker, unlidded plates ≤14 mm
For Use With (Application) Storage
Includes 2 stacks of 50 microplates
Quantity 100 Microplates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.