Learn More
Agilent BioTek BioStack Automated Microplate Stackers



BioTek BioStack microplate stacker is a compact and versatile microplate stacker, compatible with Agilent BioTek washers, dispensers, plate readers, and imaging systems.
$23102.46
Specifications
Operating Temperature Range (Metric) | 18 to 40°C |
---|---|
Hertz | 50/60 Hz |
Voltage | 100/240 V |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Humidity | 10 to 85% |
Includes: One instrument integration kit, communication cable, operator manual, microplate pedestal for storage stack loading and unloading, maintenance kit.
Catalog Number | Mfr. No. | Product Type | Plate Type | Holds | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Plate Type | Holds | Price | Quantity & Availability | |||
11-120-601
![]() ![]() |
Agilent BioTek
BSTCK2WRSN |
BioStack Microplate Stacker | ANSI/SBS Standard and low profile 96-, 384- and 1536-well plates, Maximum plate height 14.6 mm | 10-, 30- and 50-plate stacks are removable and interchangeable; 96-/384-well plates: Up to 50 plates; 1536-well plates: Up to 75 plates | N/A | N/A | |||
This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or send email to cs.webemail@thermofisher.com to discuss your specific assay requirements. | |||||||||
BTBIOSTACK4
![]() ![]() |
Agilent BioTek
BSTCK4SN |
BioStack 4 Microplate Stacker | ANSI/SBS Standard and low profile 96-, 384- and 1536-well plates; 24- and 48-well plates (model dependent) Maximum plate height: 23.2 mm | 10 and 30 plate stacks are removable and interchangeable (50-plate stacks may be used with non-lidded plates only); 96-/384-well plates: Up to 30 plates (with lids); 1536-well plates: Up to 75 plates |
Save 24% Reg : |
|
|||
BIOSTACK3WR
![]() ![]() |
Agilent BioTek
BSTCK3WRSN |
BioStack 3 Microplate Stacker | ANSI/SBS Standard and low profile 96-, 384- and 1536-well plates, Maximum plate height 14.6 mm | 10-, 30- and 50-plate stacks are removable and interchangeable; 96-/384-well plates: Up to 50 plates; 1536-well plates: Up to 75 plates | N/A | N/A | |||
This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or send email to cs.webemail@thermofisher.com to discuss your specific assay requirements. | |||||||||
BIOSTACKNEO
![]() ![]() |
Agilent BioTek
BSTCKN2SN |
BioStack NEO Microplate Stacker | ANSI/SBS Standard and low profile 96-, 384- and 1536-well plates, Maximum plate height 14.6 mm | 10-, 30- and 50-plate stacks are removable and interchangeable; 96-/384-well plates: Up to 50 plates; 1536-well plates: Up to 75 plates | N/A | N/A | |||
This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or send email to cs.webemail@thermofisher.com to discuss your specific assay requirements. | |||||||||
Description
BioTek BioStack microplate stacker is a compact and versatile microplate stacker, compatible with Agilent BioTek washers, dispensers, plate readers, and imaging systems. The rapid plate exchange speeds deliver increased throughput and enhanced efficiency and greater productivity
BioStack can accommodate assay workflows for 24- to 1536-well plates. Microscope slides can be stacked for imaging using BioTek's custom slide holders. The BioTek BioStack 4 is a delidding microplate stacker, automating the handling of lidded microplates for sensitive cell-based assays, for complete walkaway automation.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Certifications
CE marked; ETL listed
Specifications
18 to 40°C | |
100/240 V | |
10 to 85% | |
LHC v 2.12 for liquid handling instruments (optional); Gen5 2.01 for readers | |
11.3 kg |
50/60 Hz | |
CE and TUV marked. RoHS compliant. | |
One instrument integration kit; Communication cable; Operator manual; Microplate pedestal for storage stack loading and unloading; Maintenance kit | |
<25 lb. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.