Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corporation BioSuite C18 Column

Catalog No. 50781116 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
35 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
2 mm
2.1 mm
4.6 mm
21.5 mm
Particle Size:
2.5 μm
3 μm
3.5 μm
7 μm
13 μm
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Particle Size
50-781-116 250 mm 4.6 mm 3.5 μm
50-780-964 150 mm 4.6 mm 7 μm
50-781-103 150 mm 2.1 mm 3 μm
50-780-963 150 mm 2 mm 7 μm
50-780-965 150 mm 21.5 mm 13 μm
50-780-962 35 mm 4.6 mm 2.5 μm
50-781-101 50 mm 2.1 mm 3 μm
50-781-102 100 mm 2.1 mm 3 μm
50-781-104 250 mm 2.1 mm 3 μm
50-781-105 50 mm 4.6 mm 3 μm
50-781-106 100 mm 4.6 mm 3 μm
50-781-107 150 mm 4.6 mm 3 μm
50-781-108 250 mm 4.6 mm 3 μm
50-781-109 50 mm 2.1 mm 3.5 μm
50-781-110 100 mm 2.1 mm 3.5 μm
50-781-111 150 mm 2.1 mm 3.5 μm
50-781-112 250 mm 2.1 mm 3.5 μm
50-781-113 50 mm 4.6 mm 3.5 μm
50-781-114 100 mm 4.6 mm 3.5 μm
50-781-115 150 mm 4.6 mm 3.5 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Catalog No. 50-781-116 Supplier Waters Corporation Supplier No. 186002440
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BioSuite™ Peptide Analysis columns consist of two premier reversed-phase column chemistries specifically optimized for peptide mapping from simple to complicated digests.

  • BioSuite PA Columns are available in various configurations for LC or LC/MS applications, including 250 mm lengths for ultra high resolution.
  • These BioSuite™ PA-A and PA-B columns provide excellent batch-to-batch reproducibility for consistent results and are uniquely QC tested specifically for peptide mapping.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 250 mm
Max. Pressure 6000 psi (415 Bar)
Particle Shape Spherical
Particle Size 3.5 μm
Pore Size 300 Å
Quantity 1/Ea.
USP Type L1
Endcapped Yes
For Use With (Application) Peptides
Product Type Column
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.