Learn More
Waters Corporation BioSuite Cation-Exchange Column
Shop All Waters Chromatography Solutions Products
Description
- The availability of four separation chemistries provides chromatographers with the flexibility required to develop methods that separate proteins and / or peptides based upon minor charge differences.
- Nonporous (NP) and porous IEX columns are also available. Superior chromatographic resolution is possible using the nonporous IEX offerings however, greater binding capacity is obtained with the porous selections.
- In addition, selected BioSuite ion-exchange columns are available in PEEK hardware as well as in 21.5 mm preparative column sizes.
Specifications
Specifications
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 35 mm |
| Particle Shape | Spherical |
| Particle Size | 2.5 μm |
| Quantity | 1/Ea. |
| For Use With (Application) | Protein |
| Product Type | Column |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.