Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corporation BioSuite DEAE Anion-Exchange Column

Catalog No. 50780991 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
75 mm
150 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
50-780-991 150 mm
50-780-990 75 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-780-991 Supplier Waters Corporation Supplier No. 186002181
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Waters BioSuite ion-exchange column offerings include strong and weak, cation (CXC) and anion-exchangers (AXC) bonded to a pH stable, methacrylic ester-based polymeric resin.

  • The availability of four separation chemistries provides chromatographers with the flexibility required to develop methods that separate proteins and / or peptides based upon minor charge differences.
  • Nonporous (NP) and porous IEX columns are also available. Superior chromatographic resolution is possible using the nonporous IEX offerings however, greater binding capacity is obtained with the porous selections.
  • In addition, selected BioSuite ion-exchange columns are available in PEEK hardware as well as in 21.5 mm preparative column sizes.

Specifications

Diameter (Metric) 21.5 mm
Length (Metric) 150 mm
Particle Shape Spherical
Particle Size 13 μm
Pore Size 1000 Å
Quantity 1/Ea.
USP Type L23
For Use With (Application) Protein
Product Type Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.