Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corporation BioSuite Phenyl Column

Catalog No. 50780970 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
75 mm
150 mm
Diameter (Metric):
2 mm
4.6 mm
7.5 mm
21.5 mm
Product Type:
Column
Reversed Phase Column
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Product Type
50-780-970 150 mm 21.5 mm Column
50-780-966 75 mm 2 mm Column
50-780-967 75 mm 4.6 mm Column
50-780-968 150 mm 21.5 mm Reversed Phase Column
50-780-969 75 mm 7.5 mm Column
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 50-780-970 Supplier Waters Corporation Supplier No. 186002160
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Waters BioSuite™ Phenyl HIC columns provide a viable separation alternative to RPC.

  • The separation of proteins and peptides based upon hydrophobic characteristics is a powerful chromatographic technique. However, some proteins denature at elevated organic solvent concentrations making reversed-phase chromatography (RPC) difficult.
  • Waters BioSuite™ Phenyl HIC columns provide a viable separation alternative to RPC. HIC is characterized by the adsorption of compounds to a weakly hydrophobic surface at high salt concentrations, followed by elution with a decreasing salt gradient.
  • HIC combines the non-denaturing characteristics of salt precipitation with the precision of HPLC to yield excellent separation of biologically active material.

Specifications

Diameter (Metric) 21.5 mm
Length (Metric) 150 mm
Particle Shape Spherical
Particle Size 13 μm
Pore Size 1000 Å
Quantity 1/Ea.
USP Type L11
Endcapped Yes
For Use With (Application) Protein
Product Type Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.