Learn More
Honeywell Analytics™ Biosystems MultiPro Sensors
Supplier: Honeywell Analytics™ 544990
Requires : Biosystems MultiPro Multi-Gas Detector for LEL And Oxygen (Cat. No. 5448300A)
Description
- Field replaceable
- Sensor span reserve indicators for predictive maintenance
Specifications
Oxygen | |
Gas Sensor | |
0 to 30% | |
0.1% |