Learn More
Agilent BioTek Microplate Washer Flat Magnets
GSA_VAGREENER_CHOICE
For insertion into washer biomagnetic separation carriers
$2121.00 - $2939.27
Specifications
For Use With (Equipment) 405 LS Microplate Washer, 405 Touch Microplate Washer, EL406 Microplate Washer, ELx 405 Microplate Washer, ELx 50 Microplate Strip Washer
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
BT7103016
GSA_VAGREENER_CHOICE
View Documents
Agilent BioTek
7103016
96-Well Flat Magnet
Each for $2,121.00
Only null left
Add to Cart
BT7103017
GREENER_CHOICE
View Documents
Agilent BioTek
7103017
384-Well Flat Magnet
Each for $2,939.27
Only null left
Add to Cart
Description
Description
In flat-bottom well, beads pulled to band across well bottom. In round-bottom well, beads pulled to button at well bottom.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
For use with: 405 LS , 405 Touch, EL406, ELx405, ELx50 washers
Specifications
Specifications
405 LS Microplate Washer, 405 Touch Microplate Washer, EL406 Microplate Washer, ELx 405 Microplate Washer, ELx 50 Microplate Strip Washer
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.