Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotin Antibody (1D4-C5), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323510JF549
Description
Biotin Monoclonal antibody specifically detects Biotin in Nonspecies specific samples. It is validated for Flow Cytometry, ELISASpecifications
Biotin | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Non-species specific | |
Purified |
Flow Cytometry, ELISA | |
1D4-C5 | |
Flow Cytometry, ELISA | |
Vitamin B4 | |
Biotinylated Limulus hemocyanin | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction