Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotin Antibody (BK-1/39), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250120AF488
Description
Biotin Monoclonal specifically detects Biotin in Human samples. It is validated for Immunohistochemistry.Specifications
Biotin | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
Cancer, Cellular Signaling, Epitope Tags, metabolism | |
Human | |
IgG1 |
Immunohistochemistry | |
BK-1/39 | |
Immunohistochemistry | |
Vitamin B4 | |
Biotin conjugated to keyhole limpet haemocyanin | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction