Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotin Antibody (BK-1/39) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
| Antigen | Biotin |
|---|---|
| Clone | BK-1/39 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:10 - 1:500 |
| Applications | Immunohistochemistry |
Description
Biotin Monoclonal antibody specifically detects Biotin in Human samples. It is validated for ImmunohistochemistrySpecifications
| Biotin | |
| 1 mg/mL | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Signaling, Epitope Tags, metabolism | |
| PBS | |
| Biotin conjugated to keyhole limpet haemocyanin | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| BK-1/39 | |
| Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Vitamin B4 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title