Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotin Antibody (BTN/8672R), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320861MFV500
Description
Biotin Monoclonal antibody specifically detects Biotin in All samples. It is validated for Flow CytometrySpecifications
Biotin | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
All species | |
Purified |
Flow Cytometry | |
BTN/8672R | |
Flow Cytometry | |
Vitamin B4 | |
Biotinylated sheep immunoglobulin | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction