Learn More
Description
Specifications
Specifications
| Antigen | Biotin |
| Applications | Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Simple Western, Western Blot |
| Classification | Monoclonal |
| Clone | 2758A |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/mL, Simple Western 20 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 1-3 ug/mL, Immunocytochemistry 1-8 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. |
| Gene Alias | Vitamin B4 |
| Host Species | Rabbit |
| Immunogen | Biotin compound |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

