Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotin Monoclonal Antibody (BTN.4), Invitrogen™

Catalog No. PIMA511251
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA511251 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA511251 Supplier Thermo Scientific Supplier No. MA511251

Mouse Monoclonal Antibody

MA5-11251 targets Biotin in Western blot, immunomicroscopy, in situ hybridization and flow cytometry applications and shows reactivity with All samples. The MA5-11251 immunogen is biotin conjugated to BSA.

Biotin Monoclonal antibody specifically detects Biotin in Chemical samples. It is validated for Flow Cytometry, Immunomicroscopy, In Situ Hybridization (ISH), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Biotin
Applications Flow Cytometry, Immunomicroscopy, In Situ Hybridization (ISH), Western Blot
Classification Monoclonal
Clone BTN.4
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene Alias Vitamin B4
Host Species Mouse
Immunogen Biotin conjugated to BSA
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.