Learn More
Description
This product is a replacement for R&D Systems product 4670-500-01 .
131 μg is supplied as 500 μl of a 0.25 mM solution in water.
Specifications
Specifications
| Chemical Name or Material | β-Nicotinamide-N6-[2-[[6-[biotinyl]amino]hexyl]amino]-2-oxoethyl]adenine dinucleotide |
| CAS | 146385-37-7 |
| Quantity | 131 μg |
| Molecular Formula | C39H57N11O17P2S |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

