Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotin-PEG2-Amine 98.0+%, TCI America™

Click to view available options
Quantity:
25 mg
100 mg
Specifications
Specifications
Color | White |
Quantity | 100 mg |
Formula Weight | 374.50 |
Percent Purity | ≥98.0% (HPLC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | Biotin-PEG2-Amine |
Safety and Handling
TSCA | No |
Recommended Storage | Freezer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction