Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Biotin-PEG3-Azide
GREENER_CHOICESDP

Catalog No. 752450 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-245-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 75-245-0 Supplier R&D Systems™ Supplier No. 7524/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Biotin with PEG linker and reactive click handle

Biotin-PEG3-Azide is a biotinylation reagent for labeling alkyne-containing biomolecules either via Cu(I)-catalyzed Azide-Alkyne Click Chemistry reaction (CuAAC) or via Cu(I)-free Strain-Promoted Alkyne-Azide Click Chemistry (SPAAC) reaction with cyclooctyne derivatives.

Specifications

Chemical Name or Material (3 AS,4 S,6 aR)-N-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethyl]hexahydro-2-oxo-1 H-thieno[3,4-d]imidazole-4-pentanamide
CAS 875770-34-6
Quantity 50 mg
Product Line Tocris
Target General Laboratory Research Reagent
Molecular Formula C18H32N6O5S
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.