Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Tocotrienol, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014773 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4773 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4773 Supplier MilliporeSigma Supelco Supplier No. 07205100MG
Only null left
Add to Cart
Add to Cart

α-Tocotrienol is one of the isomers of vitamin E.

Featured Industries: Vitamins, Nutraceuticals and Natural Products
TRUSTED_SUSTAINABILITY

Specifications

CAS 58864-81-6
Physical Form Viscous Liquid
Linear Formula C29H44O2
MDL Number MFCD11045308
Synonym D-alpha-Tocotrienol; (R)-alpha-Tocotrienol; [R-(E,E)]-3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyl-3,7,11-tridecatrienyl)-2 H-1-benzopyran-6-ol
Type Analytical Standard
Quantity 100 mg
Formula Weight 424.66
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.