Learn More
Biotin Polyclonal Antibody, FITC, Rockland™

Rabbit Polyclonal Antibody
$174.73 - $4093.71
Specifications
Antigen | Biotin |
---|---|
Concentration | 1 mg/mL |
Applications | Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1074
![]() ![]() |
Rockland Immunochemicals
20042980100 |
100 μg |
Each for $574.79
|
|
|||||
50-199-1075
![]() ![]() |
Rockland Immunochemicals
2004298S |
25 μL |
Each for $174.73
|
|
|||||
50-199-1073
![]() ![]() |
Rockland Immunochemicals
2004298 |
20 mg |
Each for $4,093.71
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Biotin Antibody Fluorescein Conjugated is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-fluorescein, anti-Rabbit Serum, Biotin conjugated IgG and Biotin conjugated Albumin.
Biotin is a water-soluble B complex vitamin, generally classified as Vitamin B7. It is required by all organisms and can only be synthesized by bacteria, yeasts, molds, algae, and some plant species. Biotin is required for incorporation of carbon dioxide into organic compounds and exhibits a MW of ∼244 Da.Specifications
Biotin | |
Immunofluorescence, Western Blot | |
FITC | |
Rabbit | |
0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% sodium azide; pH 7.2 | |
Biotin conjugated Keyhole Limpet Hemocyanin (KLH) | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
1 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
Vitamin B4 | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.