Learn More
Invitrogen™ Biotin Polyclonal Antibody

Description
The activity measured by a solid phase binding assay is 99%. Reconstitute using 1 mL of distilled water; product may be stored for several weeks as an undiluted liquid. After dilution, do not use for more than one day. After one year, the reconstituted liquid product should be stored frozen in small undiluted aliquots, avoiding freeze/thaw cycles.
Specifications
Specifications
| Antigen | Biotin |
| Applications | ELISA, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | PBS with 20mg/mL sucrose and 0.08% sodium azide; pH 7.8 |
| Gene Alias | Vitamin B4 |
| Host Species | Goat |
| Immunogen | Biotin. |
| Purification Method | Affinity chromatography |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.