Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotin, Powder, FCC, 97.5-100.5%, Spectrum™ Chemical
SDP

Catalog No. p-7157093
Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
100 g
500 g
Packaging:
Amber Glass Bottle

BI115, 58-85-5, C10H16N2O3S

Spectrum™ Chemical Biotin, Powder, FCC is often recommended as a dietary supplement for strenghening hair and nails. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 58-85-5
Molecular Formula C10H16N2O3S
Molecular Weight (g/mol) 244.31
MDL Number MFCD00005541
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
IUPAC Name 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12

Specifications

CAS Max % 100%
Melting Range 229-232°C
Packaging Amber Glass Bottle
Quantity 500 mg
Optical Rotation +89° to +93°
Percent Purity 97.5 to 100.5%
Grade FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.