Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotin, Powder, USP, 97.5-102%, Spectrum™ Chemical
SDP

Catalog No. 18600991 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-991 5 g Amber Glass Bottle
18-600-989 500 mg Amber Glass Bottle
18-600-992 25 g Amber Glass Bottle
18-600-993 100 g Amber Glass Bottle
18-600-994 500 g Amber Glass Bottle
18-600-995 1 kg Amber Glass Bottle
18-600-990 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-600-991 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B11035GM
Add to Cart
Add to Cart

B1103, 58-85-5, C10H16N2O3S

Spectrum™ Chemical Biotin, Powder, USP is used for preventing and treating biotin deficiency. Spectrum Chemical manufactured USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 58-85-5
Molecular Formula C10H16N2O3S
Molecular Weight (g/mol) 244.31
MDL Number MFCD00005541
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
IUPAC Name 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 5 g
Specific Rotation +89° to +93°
Percent Purity 97.5 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Biotin Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.