Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotin (Vitamin B7 or Vitamin H) Antibody (BTN/2032R), Alexa Fluor™ 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP261393AF405
Description
Biotin (Vitamin B7 or Vitamin H) Monoclonal specifically detects Biotin (Vitamin B7 or Vitamin H) in All Species samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Biotin (Vitamin B7 or Vitamin H) | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Rabbit | |
Protein A purified | |
RUO | |
It recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. This monoclonal antibody is highly specific to biotin and shows no cross-reaction with other structurally related compounds. It has a very high affinity for biotin and is excellent for use in various amplification techniques. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
BTN/2032R | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
58-85-5, coenzyme R, d-biotin, Vitamin B7, vitamin H | |
Biotinylated sheep immunoglobulin | |
0.1 mL | |
Primary | |
All species | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction