Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotin (Vitamin B7 or Vitamin H) Antibody (BTN/36), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314093PCP
This item is not returnable.
View return policy
Description
Biotin (Vitamin B7 or Vitamin H) Monoclonal antibody specifically detects Biotin (Vitamin B7 or Vitamin H) in All species samples. It is validated for Flow CytometrySpecifications
| Biotin (Vitamin B7 or Vitamin H) | |
| Monoclonal | |
| PerCP | |
| 58-85-5, coenzyme R, d-biotin, Vitamin B7, vitamin H | |
| Biotinylated sheep immunoglobulin | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| BTN/36 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| All species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction