Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotinidase/BTD Antibody (006), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290315APC
Description
Biotinidase/BTD Monoclonal antibody specifically detects Biotinidase/BTD in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
Biotinidase/BTD | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry (Paraffin) | |
006 | |
ELISA, Immunohistochemistry-Paraffin | |
biotinase, biotinidase, EC 3.5.1, EC 3.5.1.12 | |
Rabbit immunized with purified, recombinant Human Biotinidase/BTD (Uniprot#: P43251, Met1-Asp543) | |
0.1 mL | |
Signal Transduction | |
686 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction