Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotinidase/BTD Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297093PE
Description
Biotinidase/BTD Polyclonal antibody specifically detects Biotinidase/BTD in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)Specifications
Biotinidase/BTD | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry-Paraffin | |
biotinase, biotinidase, EC 3.5.1, EC 3.5.1.12 | |
Produced in rabbits immunized with purified, recombinant Human Biotinidase/BTD (Uniprot#: P43251; Met1-Asp543). | |
0.1 mL | |
Signal Transduction | |
686 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction