Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Biotinyl Tyramide
GREENER_CHOICESDP

Catalog No. 624125 Shop All Novus Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-412-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 62-412-5 Supplier R&D Systems™ Supplier No. 6241/25

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Reagent widely used for signal amplification in IHC and FISH

Biotinyl Tyramide is a reagent widely used for signal amplification in IHC and fluorescent in situ hybridization (ISH). HRP catalyzes localized deposition of multiple biotinyl tyramide molecules (catalyzed reporter deposition, CARD), which can be detected using a labeled streptavidin conjugate. The localization of deposition around HRP leads to good resolution. Suitable for either fluorescence or chromogenic detection.

Specifications

Chemical Name or Material (3 AS,4 S,6 aR)-Hexahydro-N-[2-(4-hydroxyphenyl)ethyl]-2-oxo-1 H-thieno[3,4-d]imidazole-4-pentanamide
CAS 41994-02-9
Synonym (3aS,4S,6aR)-Hexahydro-N-[2-(4-hydroxyphenyl)ethyl]-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide
Quantity 25 mg
Product Line Tocris
Target Tyramide Signaling Amplification/TSA
Molecular Formula C18H25N3O3S
Formula Weight 363.47
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.