Description
Includes
- Highly specific to the azido group.
- Can be linked to the azido group through click chemistry.
- Versatile for detection through the biotin moiety.
Specifications
Specifications
| Quantity | 500 μg |
| Product Line | R&D Systems |
| Molecular Formula | C19H31N3O5S |
| Formula Weight | 413.53 Da |