Description
- Pair with muscovite mica to demonstrate effects of slight variations in a mineral's chemistry
- Useful when teaching lessons about physical properties of minerals, as well as Bowen's reaction series. Couple samples can also act as polarizers
- Useful when teaching students about variable hardness and cleavage
- Mica's thinly layered structure is unique
- Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Specifications
| Product Type | Mineral |
| For Use With (Application) | Study on variable hardness and cleavage |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 12/Pk. |
| Specimen Size | 2 to 3 cm |
| Specimen Type | Biotite Mica |