Learn More
Lonza™ BioWhittaker™ X-VIVO™ 15 Hematopoietic Serum-Free Culture Media
Catalog No. p-4895342
Click to view available options
With Additives:
Gentamicin, Phenol Red
Without Additives:
None
This item is not returnable. View return policy
Catalog No. BW04-418Q
Supplier: Lonza™ 04418Q
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
For adoptive immunotherapy and cellular therapy research. BioWhittaker™ X-VIVO™ 15 Hematopoietic Serum-Free Culture Media supports the proliferation of lymphocytes, including tumor infiltrating lymphocytes (TIL) and macrophages under serum-free conditions.
  • Does not contain any lymphokines required for activation of human lymphocytes
  • Intended for in vitro procedures
  • Device Master Records (DMRs) for these products are held in an FDA Master File
  • Chemically defined media optimized for specific cell types
  • For further manufacturing or laboratory use
  • Supports the proliferation of lymphocytes, including tumor infiltrating lymphocytes (TIL) and macrophages under serum-free conditions
  • Limited by Federal (USA) law to investigational use
  • These products are for research use only
  • Not approved for human or veterinary use, for application to humans or animals, or for use in clinical or in vitro procedures
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stem Cell Serum Free Media (SFM)
With Additives Gentamicin, Phenol Red
Without Additives None
Quantity 1 L
Content And Storage 2-8°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.