Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biphenyl-2-boronic acid, +97%, Thermo Scientific™

Catalog No. AC394880050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4688-76-0
Molecular Formula C12H11BO2
Molecular Weight (g/mol) 198.028
MDL Number MFCD00136929
InChI Key HYCYKHYFIWHGEX-UHFFFAOYSA-N
Synonym 2-biphenylboronic acid, biphenyl-2-boronic acid, biphenyl-2-ylboronic acid, 2-biphenylboronicacid, 2-phenylphenyl boranediol, 2-phenylphenyl boronic acid, 1,1'-biphenyl-2-ylboronic acid, 2-biphenylylboronic acid, 1,1'-biphenylboronic acid, 2-phenylbenzeneboronic acid
PubChem CID 4589187
IUPAC Name (2-phenylphenyl)boronic acid
SMILES B(C1=CC=CC=C1C2=CC=CC=C2)(O)O

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Linear Formula C6H5C6H4B(OH)2
Quantity 5 g
Formula Weight 198.05
Percent Purity ≥96.0% (HPLC)
Physical Form Powder or Low Melting Solid
Chemical Name or Material Biphenyl-2-boronic acid
Show More Show Less
Hazard Statement GHS P Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If skin irritation or rash occurs: Get medical advice/attention. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.