Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biphenyl-3,3',5,5'-tetracarboxylic acid, Thermo Scientific Chemicals

Catalog No. AC465830010
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465830010 1 g
AC465831000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465830010 Supplier Thermo Scientific Chemicals Supplier No. 465830010
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4371-28-2
Molecular Formula C16H10O8
Molecular Weight (g/mol) 330.25
MDL Number MFCD16621475
InChI Key QURGMSIQFRADOZ-UHFFFAOYSA-N
IUPAC Name [1,1'-biphenyl]-3,3',5,5'-tetracarboxylic acid
SMILES OC(=O)C1=CC(=CC(=C1)C(O)=O)C1=CC(=CC(=C1)C(O)=O)C(O)=O

Specifications

Alpha Vector BIPHENYLTETRACARBOXYLICACID
Melting Point >400°C
Color Brown to White
Infrared Spectrum Conforms
Assay Percent Range 94.0 to 106.0 % (Titration with NaOH)
Packaging glass bottle
Quantity 1 g
UN Number 3077
Formula Weight 330.25
Physical Form Crystalline Solid or Powder
Chemical Name or Material Biphenyl-3,3 ',5,5 '-tetracarboxylic acid
Show More Show Less
Hazard Category GHS07: Harmful
Hazard Statement GHS09: Environmental hazard
ShelfLife 5 years
storageNote1 normal conditions

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.