Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biphenyl-4,4'-diboronic acid bis(pinacol) ester, 95%

Catalog No. p-7034286
Click to view available options
Quantity:
1 g
5 g

CAS: 207611-87-8 | C24H32B2O4 | 406.14 g/mol

Biphenyl-4,4'-diboronic acid bis(pinacol) ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl-4,4′-diboronic acid bis(pinacol) ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207611-87-8
Molecular Formula C24H32B2O4
Molecular Weight (g/mol) 406.14
MDL Number MFCD08669545
InChI Key INNVTCAYPFZCAL-UHFFFAOYSA-N
Synonym 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl, 4,4'-biphenyldiboronic acid dipinacol ester, 4,4'-biphenyldiboronic acid pinacol ester, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl biphenyl, biphenyl-4,4'-diboronic acid bis pinacol ester, 4,4'-biphenyldiboronic acid bis pinacol ester, 4,4,5,5-tetramethyl-2-4'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl-4-yl-1,3,2-dioxaborolane, amtb257, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl biphenyl
PubChem CID 10525359
IUPAC Name 4,4,5,5-tetramethyl-2-[4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]phenyl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 406.14
Percent Purity 95%
Chemical Name or Material Biphenyl-4,4'-diboronic acid bis(pinacol) ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.