Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biphenyl-4-carboxaldehyde 95.0+%, TCI America™
SDP

Catalog No. B024225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B024225G 25 g
B02425G 5 g
B0242250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B024225G Supplier TCI America Supplier No. B024225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3218-36-8
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.22
MDL Number MFCD00006947
InChI Key ISDBWOPVZKNQDW-UHFFFAOYSA-N
Synonym 4-biphenylcarboxaldehyde, 4-biphenylaldehyde, p-phenylbenzaldehyde, 1,1'-biphenyl-4-carbaldehyde, biphenyl-4-carboxaldehyde, 4-formylbiphenyl, p-biphenylylaldehyde, 1,1'-biphenyl-4-carboxaldehyde, p-biphenylaldehyde, p-biphenylcarboxaldehyde
PubChem CID 76689
IUPAC Name [1,1'-biphenyl]-4-carbaldehyde
SMILES O=CC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 58°C
Boiling Point 152°C
Quantity 25 g
Formula Weight 182.22
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Biphenyl-4-carboxaldehyde
EINECSNumber (4)-1745
RTECSNumber DV1767000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.