Alfa Aesar™ Biphenyl-4-carboxylic acid, 98%

Manufacturer: Alfa Aesar™ A1243814

Quantity 25g

 View more versions of this product

Catalog No. AAA1243814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 92-92-2
    Molecular Formula C13H10O2
    Formula Weight 198.22
    MDL Number MFCD00002553
    Beilstein 973519
    Synonym 4-Phenylbenzoic acid
    Chemical Name or Material Biphenyl-4-carboxylic acid

    Specifications

    Formula Weight 198.22
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00002553
    Beilstein 973519
    Melting Point 223° to 227°C
    Synonym 4-Phenylbenzoic acid
    Quantity 25g
    Health Hazard 1 Warning
    RTECS Number DV1925100
    CAS 92-92-2
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Odor Odorless
    TSCA TSCA
    Molecular Formula C13H10O2
    Chemical Name or Material Biphenyl-4-carboxylic acid
    Assay 98%

    Description

    Applications
    Biphenyl-4-carboxylic acid is employed in the synthesis, characterization of europium, terbium complexes.

    Solubility
    Insoluble in water.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit