Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biphenylene, 99+%, Thermo Scientific Chemicals

Catalog No. p-67156
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 259-79-0
Molecular Formula C12H8
Molecular Weight (g/mol) 152.2
MDL Number MFCD00001110
InChI Key IFVTZJHWGZSXFD-UHFFFAOYSA-N
Synonym diphenylene, cyclobutadibenzene, dibenzocyclobutadiene, 1,1'-biphenylene, unii-0z64i7d5m2, 8ci 9ci, biphenylen, biphenylene, biphenylene, 99+%, pubchem9106
PubChem CID 9214
ChEBI CHEBI:33079
IUPAC Name biphenylene
SMILES C1=CC=C2C(=C1)C3=CC=CC=C23

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 109°C to 112°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 250 mg
Beilstein 05, 298
Solubility Information Solubility in water: 9.60mg/l (calc). Other solubilities: organic solvents general
Formula Weight 152.2
Percent Purity 99+%
Physical Form Needles
Chemical Name or Material Biphenylene, 99+%
Show More Show Less
Hazard Statement GHS H Statement:
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.