Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BirA Antibody (6C4c7), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259938AF350
Description
BirA Monoclonal specifically detects BirA in E. coli, Yeast samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
BirA | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
birA | |
GST fused to E.coli BirA R118G | |
Epitope Tags | |
948469 | |
E. coli | |
IgG1 κ |
Western Blot, Immunocytochemistry, Immunofluorescence | |
6C4c7 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Bifunctional protein birA, Bio R, BioR, Biotin [acetyl CoA carboxylase] synthetase | |
Mouse | |
0.1 mL | |
Primary | |
Specificity of e. coli BirA antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction