Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BirA Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299832AF532
Description
BirA Polyclonal specifically detects BirA in E. coli samples. It is validated for ELISA.Specifications
BirA | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
E. coli | |
Purified |
ELISA | |
Alexa Fluor 532 | |
Bifunctional protein birA, Bio R, BioR, Biotin [acetyl CoA carboxylase] synthetase | |
Produced in rabbits immunized with purified, recombinant BirA (P06709; Met1-Lys321). | |
0.1 mL | |
Epitope Tags | |
948469 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction