Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BIRB 796
GREENER_CHOICE

Catalog No. 598910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-891-0 10 mg
59-895-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 59-891-0 Supplier Tocris Bioscience™ Supplier No. 5989/10
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

High affinity and selective p38 kinase inhibitor

BIRB 796 is a high affinity and selective p38 kinase inhibitor (Kd = 50-100 pM). Exhibits no significant inhibition on a panel of related kinases. Inhibits LPS-induced TNFα production in human PBMCs and whole blood (IC50 values are 21 and 960 nM, respectively). Also inhibits JNK2α2 and c-Raf-1 (IC50 values are 98 nM and 1.4 μM, respectively). Cell permeable.

Specifications

Chemical Name or Material N-[3-(1,1-Dimethylethyl)-1-(4-methylphenyl)-1H-pyrazol-5-yl]-N′-[4-[2-(4-morpholinyl)ethoxy]-1-naphthalenyl]urea
CAS 285983-48-4
Synonym Doramapimod
Quantity 10 mg
Target p38 Inhibitors
Molecular Formula C31H37N5O3
Formula Weight 527.66
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.