Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Birch pollen Bet V1 Antibody (BV16), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281093G
Description
Birch pollen Bet V1 Monoclonal specifically detects Birch pollen Bet V1 in Plant samples. It is validated for Western Blot, ELISA.Specifications
Birch pollen Bet V1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
This antibody binds to Birch pollen Bet V1. | |
Store at 4°C in the dark. | |
IgG κ |
Western Blot, ELISA | |
BV16 | |
Western Blot, ELISA | |
Allergen Bet v I-A, Allergen=Bet v 1-A, BETVI, BETVIA, Major pollen allergen Bet v 1-A | |
Birch pollen Bet V1-A, a major allergen of the European white birch. | |
0.1 mL | |
Primary | |
Plant | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction