Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BIRT 377
GREENER_CHOICE

Catalog No. 477610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-761-0 10 mg
47-765-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-761-0 Supplier Tocris Bioscience™ Supplier No. 4776/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent negative allosteric modulator of LFA-1

BIRT 377 is a potent negative allosteric modulator of LFA-1 (lymphocyte function-associated antigen-1) (Kd = 26 nM). BIRT 377 binds to the I-domain of LFA-1. BIRT 377 reversibly inhibits LFA-1 mediated binding of SKW3 leukemia cells to ICAM-1. BIRT 377 inhibits superantigen (SEB)-induced IL-2 production from lymphocytes in vitro and attenuates the SEB-induced increase in IL-2 plasma levels in a mouse model in vivo. Orally available.

Specifications

Quantity 10 mg
Product Type BIRT 377
Purity >98%
Molecular Weight (g/mol) 442.13
Inhibitors BIRT 377

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.