Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2,2,2-trifluoroethyl) Carbonate 98.0+%, TCI America™
SDP

Catalog No. p-7123696 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1513-87-7
Molecular Formula C5H4F6O3
Molecular Weight (g/mol) 226.074
MDL Number MFCD11857845
InChI Key WLLOZRDOFANZMZ-UHFFFAOYSA-N
Synonym Carbonic Acid Bis(2,2,2-trifluoroethyl) Ester
PubChem CID 11447463
IUPAC Name bis(2,2,2-trifluoroethyl) carbonate
SMILES C(C(F)(F)F)OC(=O)OCC(F)(F)F

Specifications

Color Yellow
Boiling Point 118°C
Quantity 5 g
UN Number 3272
Formula Weight 226.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2,2,2-trifluoroethyl) Carbonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.