Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2,2,2-trifluoroethyl) (Methoxycarbonylmethyl)phosphonate 95.0+%, TCI America™
SDP

Catalog No. B171425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B171425G 25 g
B17141G 1 g
B17145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B171425G Supplier TCI America Supplier No. B171425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88738-78-7
Molecular Formula C7H9F6O5P
Molecular Weight (g/mol) 318.108
MDL Number MFCD00009901
InChI Key PVSJXEDBEXYLML-UHFFFAOYSA-N
Synonym methyl 2-bis 2,2,2-trifluoroethoxy phosphoryl acetate, bis 2,2,2-trifluoroethyl methoxycarbonylmethyl phosphonate, methyl bis 2,2,2-trifluoroethyl phosphonoacetate, bis 2,2,2-trifluoroethyl phosphonoacetic acid methyl ester, methyl bis 2,2,2-trifluoroethoxy phosphoryl acetate, methyl p,p-bis 2,2,2-trifluoroethyl phosphonoacetate, acetic acid, bis 2,2,2-trifluoroethoxy phosphinyl-, methyl ester, acmc-209qx4
PubChem CID 184894
IUPAC Name methyl 2-[bis(2,2,2-trifluoroethoxy)phosphoryl]acetate
SMILES COC(=O)CP(=O)(OCC(F)(F)F)OCC(F)(F)F

Specifications

Color Yellow
Quantity 25 g
Formula Weight 318.11
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2,2,2-trifluoroethyl) (Methoxycarbonylmethyl)phosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.