Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2,2,2-trifluoroethyl) Phosphite 94.0+%, TCI America™
SDP

Catalog No. B262725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B262725G 25 g
B26275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B262725G Supplier TCI America Supplier No. B262725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 92466-70-1
Molecular Formula C4H4F6O3P+
Molecular Weight (g/mol) 245.037
InChI Key IMDCVAFSSZPRRM-UHFFFAOYSA-N
Synonym bis 2,2,2-trifluoroethyl phosphonate, phosphonic acid bis 2,2,2-trifluoroethyl ester, oxo-bis 2,2,2-trifluoroethoxy phosphanium, phosphonic acid, bis 2,2,2-trifluoroethyl ester, bis trifluoroethyl phosphonate, bis 2,2,2-trifluoroethyl phosphite, bis 2,2,2-trifluoroethyl hydrogen phosphite, oxobis 2,2,2-trifluoroethoxy-??-phosphanylium, bis 2,2,2-trifluoroethyl phosphite, technical grade
PubChem CID 6386648
IUPAC Name oxo-bis(2,2,2-trifluoroethoxy)phosphanium
SMILES C(C(F)(F)F)O[P+](=O)OCC(F)(F)F

Specifications

Color Yellow
Boiling Point 44°C
Quantity 25 g
Formula Weight 246.05
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2,2,2-trifluoroethyl) Phosphite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.